missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma BIOshell A160 Peptide PCS-C18 HPLC Column
BIOshell™ A160 Peptide PCS-C18 columns are specifically engineered to provide efficient separation of peptides as well as small proteins.
Supplier: MilliporeSigma 50663U
Description
These columns contain advanced Fused-Core™ particles with pores strategically sized to 160 Å to enable optimized peptide diffusion. This attribute makes these columns an excellent choice for peptide mapping.
- The unique PCS-C18 chemistry enables exceptional peak shape and enhanced loading capacity of basic peptides as compared to traditional C18 chemistries.
- In addition, the PCS-C18 facilitates the use of mobile phases with lower concentrations of mobile phase modifiers without any compromise to column performance.
- This column also provides orthogonal selectivity to ES-C18, ES-CN (cyano), and phenyl-hexyl chemistries.
Specifications
| 3.0 mm | |
| 2.7 μm | |
| Positively Charged Surface (PCS) | |
| HPLC Column |
| 150 mm | |
| 160 Å | |
| Stainless Steel | |
| HPLC |