missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma BIOshell A160 Peptide PCS-C18 HPLC Column

Catalog No. 01000911
Change view
Click to view available options
Diameter (Metric):
1.5 mm
2.1 mm
3.0 mm
4.6 mm
Length (Metric):
5 mm
50 mm
100 mm
150 mm
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Length (Metric)
01000911 4.6 mm 150 mm
01000908 4.6 mm 5 mm
01000909 3.0 mm 5 mm
01000910 2.1 mm 5 mm
01000912 4.6 mm 100 mm
01000913 4.6 mm 50 mm
01000914 3.0 mm 150 mm
01000915 3.0 mm 100 mm
01000916 3.0 mm 50 mm
01000917 2.1 mm 150 mm
01000918 2.1 mm 100 mm
01000919 2.1 mm 50 mm
01000920 1.5 mm 150 mm
01000921 1.5 mm 100 mm
01000922 1.5 mm 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 01000911 Supplier MilliporeSigma Supplier No. 50668U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BIOshell™ A160 Peptide PCS-C18 columns are specifically engineered to provide efficient separation of peptides as well as small proteins.

These columns contain advanced Fused-Core™ particles with pores strategically sized to 160 Å to enable optimized peptide diffusion. This attribute makes these columns an excellent choice for peptide mapping.

  • The unique PCS-C18 chemistry enables exceptional peak shape and enhanced loading capacity of basic peptides as compared to traditional C18 chemistries.
  • In addition, the PCS-C18 facilitates the use of mobile phases with lower concentrations of mobile phase modifiers without any compromise to column performance.
  • This column also provides orthogonal selectivity to ES-C18, ES-CN (cyano), and phenyl-hexyl chemistries.

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 150 mm
Particle Size 2.7 μm
Pore Size 160 Å
Stationary Phase PCS-C18 (dimethyloctadecylsilane, positively charged ligand)
Product Line BIOshell
Phase Positively Charged Surface (PCS)
Material Stainless Steel
Technique HPLC
Product Type HPLC Column