missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ BIOshell™ A160 Peptide CN Guard Cartridge, 2.7 μm Particle Size
BIOshell™ Guard Columns provide physical (filtration) and chemical protection for costly analytical columns without compromising the very high performance of BIOshell™ columns.
$868.00 - $868.00
Specifications
| Carbon Load | 1.5% |
|---|---|
| Quantity | 3/Pk. |
| pH | 1 to 8 |
| Particle Size | 2.7 μm |
| Length (Metric) | 0.5 cm |
Description
BIOshell™ guard columns are capable of continuous use at pressures up to 9,000 psi (600 bar) with only hand-tightening. Guard cartridges are easily replaced without removing the guard column holder from the flow path. The cartridge is packed with BIOshell™ Fused-Core particles. Order the guard column holder (66841-U) separately.Specifications
| 1.5% | |
| 1 to 8 | |
| 0.5 cm | |
| 90 m2/g | |
| Stainless Steel Hardware (Column) | |
| HPLC | |
| BIOshell |
| 3/Pk. | |
| 2.7 μm | |
| 160 Å | |
| Fused-Core HPLC Guard Cartridge | |
| Reversed Phase | |
| Yes |
BIOshell is a trademark of Sigma-Aldrich Co. LLC