missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ BIOshell™ A160 Peptide C18 HPLC Column, 160 Å, 4.7 μm Particle Size

Catalog No. 111008370
Change view
Click to view available options
Length (Metric):
3 cm
5 cm
7.5 cm
10 cm
15 cm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
111008370 3 cm 3 mm
111008369 3 cm 2.1 mm
111007926 5 cm 2.1 mm
111007669 7.5 cm 2.1 mm
111007512 10 cm 2.1 mm
111007057 15 cm 2.1 mm
111007927 5 cm 3 mm
111007513 10 cm 3 mm
111007058 15 cm 3 mm
111007928 5 cm 4.6 mm
111007514 10 cm 4.6 mm
111007059 15 cm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 111008370 Supplier MilliporeSigma™ Supelco™ Supplier No. 67007U

BIOshell™ 160 Å Peptide C18 columns are specifically engineered to provide efficient separation of peptides as well as small proteins.

BIOshell™ 160 Å Peptide C18 columns contain advanced Fused-Core particles with pores strategically sized to 160 Å to enable optimized peptide diffusion. This attribute makes these columns an excellent choice for peptide mapping. Additionally, the sterically-protected C18 ligands provide extra stability allowing the columns to be used at an extended pH range (2-9) and high temperatures (up to 90°C). This greatly expands the application range for the separation of biomolecules

Specifications

Diameter (Metric) 3 mm
Length (Metric) 3 cm
Particle Size 4.7 μm
pH 1 to 8
Pore Size 160 Å
Surface Area 60 m2/g
Stationary Phase C18 (octadecyl)
Product Line BIOshell
Matrix C18 (octadecyl) Phase/Fused-Core Particle Platform
Carbon Load 4%
Endcapped No
Phase Reversed Phase
Material Stainless Steel
Technique Fused-Core HPLC
Show More Show Less

BIOshell is a trademark of Sigma-Aldrich Co. LLC