missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ BIOshell™ A160 Peptide C18 Column, 2 μm Particle Size

Catalog No. 111007245
Change view
Click to view available options
Length (Metric):
2 cm
3 cm
5 cm
7.5 cm
10 cm
15 cm
Diameter (Metric):
2.1 mm
3 mm
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
111007245 2 cm 2.1 mm
111007246 3 cm 2.1 mm
111007247 5 cm 2.1 mm
111007122 7.5 cm 2.1 mm
111006981 10 cm 2.1 mm
111006758 15 cm 2.1 mm
111007248 2 cm 3 mm
111007249 3 cm 3 mm
111007250 5 cm 3 mm
111007123 7.5 cm 3 mm
111006982 10 cm 3 mm
111006759 15 cm 3 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 111007245 Supplier MilliporeSigma™ Supelco™ Supplier No. 67234U

Shell Thickness: 0.4 μm. Solid Core: 1.2 μm. Extent of labeling: 4.0%

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 2 cm
Particle Size 2 μm
Pore Size 160 Å
Surface Area 65 m2/g
Stationary Phase C18 (octadecyl)
Product Line BIOshell
Matrix C18 (octadecyl) Phase/Spherical Silica Particle Platform
Endcapped No
Phase Reversed Phase
Material Stainless Steel Hardware
Technique Fused-Core HPLC
Show More Show Less

BIOshell is a trademark of Sigma-Aldrich Co. LLC