missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ BIOshell™ 160 Å Peptide C18 Guard Column, 2 μm Particle Size

Catalog No. 111008442
Change view
Click to view available options
Diameter (Metric) Inner:
2.1 mm
3 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
111008442 3 mm
111008441 2.1 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111008442 Supplier MilliporeSigma™ Supelco™ Supplier No. 67282U

BIOshell™ Guard Columns provide physical (filtration) and chemical protection for costly analytical columns without compromising the very high performance of BIOshell™ columns.

BIOshell™ guard columns are capable of continuous use at pressures up to 9,000 psi (600 bar) with only hand-tightening. Guard cartridges are easily replaced without removing the guard column holder from the flow path. The cartridge is packed with BIOshell™ Fused-Core particles. Order the guard column holder (66841-U) separately.

Specifications

pH 1 to 8
Particle Size 2 μm
Length (Metric) 5 cm
Pore Size 160 Å
Surface Area 65 m2/g
Type Fused-Core HPLC Guard Cartridge
Diameter (Metric) Inner 3 mm
Material Stainless Steel Hardware
Phase Reversed Phase
For Use With (Application) HPLC
Endcapped No
Product Line BIOshell
Show More Show Less

BIOshell is a trademark of Sigma-Aldrich Co. LLC