missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Biosate™ Peptone
An animal origin (AO) mixed hydrolysate comprised of 65% pancreatic digest of casein and 35% yeast extract.
$197.16 - $197.16
Specifications
| Content And Storage | Storage conditions: 2°C to 25°C. Â Store in cool, dry conditions in well-sealed containers. |
|---|---|
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| For Use With (Application) | Nutritional supplement in vitamin assays, susceptibility testing, and other laboratory media, and microbial fermentation in which the high salt content will not interfere |
| Product Line | Biosate |
Description
The combination of pancreatic digest of casein and yeast extract provides synergistic nutritional benefits that are not provided by the components alone, resulting in a unique nutritional profile. Customers have shown the combined use of these two peptones, when supplied as Biosate Peptone, improves toxin production with Clostridial species for vaccines, such as Clostridium botulinum and Clostridium perfringens.Specifications
| Storage conditions: 2°C to 25°C. Â Store in cool, dry conditions in well-sealed containers. | |
| Dehydrated Culture Media | |
| Biosate |
| Powder | |
| Nutritional supplement in vitamin assays, susceptibility testing, and other laboratory media, and microbial fermentation in which the high salt content will not interfere |