missing translation for 'onlineSavingsMsg'
Learn More

Amsbio LLC (formerly Genlantis) BioPORTER™ Protein Delivery Reagents and Kits

Catalog No. BP502401
Encompass
Change view
Click to view available options
Product Type:
Protein Delivery Reagent
QuikEase Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
BP502401 Protein Delivery Reagent
BP502424 QuikEase Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP502401 Supplier Amsbio LLC (formerly Genlantis) Supplier No. BP502401
Only null left

Translocate functional proteins directly into a wide range of cell types

  • Efficient delivery of proteins, peptides and other macromolecules into a wide variety of cell types
  • Fast and easy protocol
  • Unique lipid formulation allows direct translocation of proteins into living cells

Specifications

Product Type Protein Delivery Reagent
Quantity 24 Tests
Content And Storage -20°C