missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BIOLiner™ Rotor Bucket Adapters
Combine these rotor bucket adapters with the BIOLiner Rotor Bucket for a complete system.
$1,516.33 - $2,225.83
| Catalog Number | Mfr. No. | Capacity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity | Price | Quantity & Availability | |||||
|
75003671
|
Thermo Scientific™
75003671 |
5/7 mL Blood Collection Tube |
Pack of 4 Each for $1,819.71
|
|
|||||
|
75003672
|
Thermo Scientific™
75003672 |
10 mL Blood Collection Tube |
Pack of 4 Each for $1,931.00
|
|
|||||
|
75003674
|
Thermo Scientific™
75003674 |
40 x 50 mL Conical Tubes |
Pack of 4 Each for $2,225.83
|
|
|||||
|
75003691
|
Thermo Scientific™
75003691 |
250 mL Corning Conical Bottles |
Pack of 4 Each for $1,516.33
|
|
|||||
Description
Combine Thermo Scientific™ BIOLiner™ Rotor Bucket Adapters with the BIOLiner Rotor Bucket for a complete system.
Ordering Information:
Individual adapters and bottle/adapter packages are offered.
- Capacity range: 3L to 980mL
- Maximum tube dimensions (O.D. x L): 98 × 124mm
Includes:
75-003-692 includes 750mL flat-bottom bottle with all round adapters for the TX-750 round bucket and four 500mL Corning™ conical bottle adapters.
Specifications
| 4 |