missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Biohazard Sign Vinyl BIOHAZARD
Identify the actual or potential presence of a biologically hazardous material using Biohazard Labels made from self-sticking vinyl (B-585) material.
$5.15 - $20.87
Specifications
| Material | Vinyl |
|---|---|
| Mounting Type | Self-adhesive |
| Shape | Rectangular |
| Languages | English |
| Color | Black on Orange |
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
|
17108797
|
Brady™
17753SLSA |
3.5 x 12 in. |
Each for $11.23
|
|
|||||
|
17108808
|
Brady™
89170A* |
3.5 x 5 in. |
Each for $5.43
|
|
|||||
|
17108810
|
Brady™
89169A |
5 x 3.5 in. |
Each for $5.15
|
|
|||||
|
17108806
|
Brady™
85553A* |
7 x 10 in. |
Each for $15.34
|
|
|||||
|
17108807
|
Brady™
85554A |
10 x 14 in. |
Each for $20.87
|
|
|||||
Description
- Good resistance to most common chemicals, oils and water
- Warning labels support compliance with OSHA 29 CFR 1910.1030 Regulation for Bloodborne Pathogens Labelling
- Warning labels support compliance with DOT 49 CFR 172.400(b) Labelling Requirements for transportation
- Service temperature of -40°F to +122°F (-40°C to +50°C)
Specifications
| Vinyl | |
| Rectangular | |
| Black on Orange |
| Self-adhesive | |
| English |