missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Biohazard Sign Vinyl BIOHAZARD
Description
- Good resistance to most common chemicals, oils and water
- Warning labels support compliance with OSHA 29 CFR 1910.1030 Regulation for Bloodborne Pathogens Labelling
- Warning labels support compliance with DOT 49 CFR 172.400(b) Labelling Requirements for transportation
- Service temperature of -40°F to +122°F (-40°C to +50°C)
Specifications
Specifications
| Material | Vinyl |
| Mounting Type | Self-adhesive |
| Shape | Rectangular |
| Languages | English |
| Dimensions (L x W) | 5 x 3.5 in. |
| Product Type | BioHazard Safety Label |
| Color | Black on Orange |