missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Biohazard Sign Vinyl BIOHAZARD
Identify the actual or potential presence of a biologically hazardous material using Biohazard Labels made from self-sticking vinyl (B-585) material.
Supplier: Brady™ 85553A*
Description
- Good resistance to most common chemicals, oils and water
- Warning labels support compliance with OSHA 29 CFR 1910.1030 Regulation for Bloodborne Pathogens Labelling
- Warning labels support compliance with DOT 49 CFR 172.400(b) Labelling Requirements for transportation
- Service temperature of -40°F to +122°F (-40°C to +50°C)
Chemical Identifiers
| Vinyl | |
| Rectangular | |
| Black on Orange |
| Self-adhesive | |
| English |
Specifications
| Vinyl | |
| Rectangular | |
| 7 x 10 in. | |
| Black on Orange |
| Self-adhesive | |
| English | |
| BioHazard Safety Label |