missing translation for 'onlineSavingsMsg'
Learn More

Biodiesel Fuel Standard, SPEX CertiPrep™

Catalog No. bd3515
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BD3515 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BD3515 Supplier SPEX CertiPrep Group LLC Supplier No. BD3515
Only null left

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Tricaprin621-71-68000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 621-71-6 , 1718-52-1 , 122-32-7 , 56-81-5
Molecular Formula C33H62O6
Molecular Weight (g/mol) 554.853
InChI Key LADGBHLMCUINGV-UHFFFAOYSA-N
PubChem CID 69310
ChEBI CHEBI:77388
IUPAC Name 2,3-di(decanoyloxy)propyl decanoate
SMILES CCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCC)OC(=O)CCCCCCCCC

Specifications

Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Min % 0.8, 99.2
CAS Max % 0.8, 99.2
Physical Form Fluid
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Pyridine
Industry Type Petrochemical Testing
Quantity 1 mL
Chemical Name or Material Biodiesel Fuel Standard in Pyridine
Show More Show Less
Hazard Category Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. Causes skin irritation. May cause drowsiness or dizziness. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure.
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only