accessibility menu, dialog, popup

UserName

CAS RN 122-32-7

IUPAC Name: 1,3-bis[(9Z)-octadec-9-enoyloxy]propan-2-yl (9Z)-octadec-9-enoate
Synonyms: triolein trioleate glycerin olein, tri- kaolube 190 trioleyl glycerol edenor nhti-G emerest 2423 trioleoylglyceride trioleate actor LO 1
Molecular Weight (g/mol): 885.45
Molecular Formula: C57H104O6
InChi Key: PHYFQTYBJUILEZ-IUPFWZBJSA-N
SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C/CCCCCCCC)OC(=O)CCCCCCC\C=C/CCCCCCCC
Molecular Weight (g/mol) 
  • (4)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (2)
  • (6)
  • (4)
  • (5)
  • (9)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)