missing translation for 'onlineSavingsMsg'
Learn More

Biochanin A 98.0+%, TCI America™

Catalog No. B4098200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4098200MG 200 mg
B40981G 1 g
B40985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B4098200MG Supplier TCI America Supplier No. B4098200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 491-80-5
Molecular Formula C16H12O5
Molecular Weight (g/mol) 284.27
MDL Number MFCD00006839
InChI Key WUADCCWRTIWANL-UHFFFAOYSA-N
Synonym biochanin a, biochanin, 4'-methylgenistein, 5,7-dihydroxy-4'-methoxyisoflavone, 5,7-dihydroxy-3-4-methoxyphenyl-4h-chromen-4-one, genistein 4-methyl ether, pratensol, biochanine a, 5,7-dihydrox-4'-methoxyisoflavone, 4h-1-benzopyran-4-one, 5,7-dihydroxy-3-4-methoxyphenyl
PubChem CID 5280373
ChEBI CHEBI:17574
IUPAC Name 5,7-dihydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one
SMILES COC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O

Specifications

Melting Point 217°C
Quantity 200 mg
Formula Weight 284.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Biochanin A
RTECSNumber DJ3002500
TSCA Yes