Learn More
Thermo Scientific™ BioBasic™ 18 HPLC Columns
Use this 300Å wide-pore silica for the separation of small to medium proteins and other biomolecules.
Supplier:  Thermo Scientific™ 72105152130
 View more versions of this product
Catalog No. 72105152130
Only null left
Explore available promotions
Description
Description
Separate small to medium peptides with Thermo Scientific™ BioBasic™ 18 LC Columns.
  • 300Å pore size for maximum performance with biomolecules
  • High peak capacity stationary phase
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
Specifications
Specifications
Analytical Column
150 mm
5 μm
300 Å
60°C
C18
Spherical, Fully Porous, Ultrapure Silica
Reversed Phase
Reversed Phase
Bio-LC
2.1 mm
5800 psi (400 bar)
2 to 9
100 m²/g
L1
BioBasic
9%
Yes
Analytical Column
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.