Learn More
Promega Bio-Glo™ Luciferase Assay System DFS Item
The Bio-Glo™ Luciferase Assay System provides a highly sensitive, robust, homogeneous reagent, the Bio-Glo™ Assay reagent with a new luciferase substrate, for the detection of firefly luciferase reporter gene expression in the ADCC Reporter Bioassay.
$416.36 - $2458.64
Specifications
Content And Storage Varies Among Components
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRG7941 DFS Item
View Documents
Promega
G7941
10 mL
Each for $416.36
Only null left
PRG7940 DFS Item
View Documents
Promega
G7940
100 mL
Each for $2,458.64
Only null left
Description
Description
  • Stable, Inhibitor-Resistant Luciferase Reagent for Bioassays
  • Brighter, longer-lasting signal enables long incubations
  • Room-temperature storage provides added convenience
Specifications
Specifications
Varies Among Components
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'