missing translation for 'onlineSavingsMsg'
Learn More

Promega Bio-Glo™ Luciferase Assay System

Catalog No. prg7940
Encompass
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG7940 100 mL
PRG7941 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRG7940 Supplier Promega Supplier No. G7940
Only null left

The Bio-Glo™ Luciferase Assay System provides a highly sensitive, robust, homogeneous reagent, the Bio-Glo™ Assay reagent with a new luciferase substrate, for the detection of firefly luciferase reporter gene expression in the ADCC Reporter Bioassay.

  • Stable, Inhibitor-Resistant Luciferase Reagent for Bioassays
  • Brighter, longer-lasting signal enables long incubations
  • Room-temperature storage provides added convenience

Specifications

Quantity 100 mL
Content And Storage Varies Among Components