missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abbott™ BinaxNOW™ Malaria Tests
Rapid malaria test cleared by the FDA
Specifications
| CPT Code | 87899 |
|---|---|
| Clia Complexity | Moderate |
| Detectable Analytes | P. vivax Antigen, P. ovale Antigen, P. malariae Antigen |
| Sample Type | Whole Blood |
| Test Time | 15 min. |
| Catalog Number | Mfr. No. | Includes | Type | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Includes | Type | Price | Quantity & Availability | ||||
BNX665010
|
Abbott™
665010 |
BinaxNOW Positive Control | BinaxNOW Positive Control |
N/A
|
|
||||
BNX665000
|
Abbott™
660000 |
Malaria Test Kit (Global), with built-in control | Malaria Test Kit (Global) |
N/A
|
|
||||
Description
A rapid and simple diagnostic test that can accurately differentiate a deadly P. falciparum infection from a pan-malarial infection caused by P. vivax, P. ovale, or P. malariae
- For differential diagnosis of a malaria infection
- Just three simple steps, using one reagent
- Reliable results in 15 minutes
- CLIA Complexity: Moderate
- CPT Code: 87899
- Sensitivity: 99.7% (P.f.); 93.5% (P.v.)
- Specificity: 94.2% (P.f.); 99.8% (P.v.)
BNX665025: Malaria Test Kit - US
BNX665010: Positive Control
Specifications
| 87899 | |
| P. vivax Antigen, P. ovale Antigen, P. malariae Antigen | |
| 15 min. |
| Moderate | |
| Whole Blood |