missing translation for 'onlineSavingsMsg'
Learn More

Abbott™ BinaxNOW™ Malaria Tests

Rapid malaria test cleared by the FDA

Supplier:  Abbott™ 665010

Encompass

 Afficher plus de versions de ce produit

Numéro de catalogue. BNX665010

Il en reste null

Description

Description

A rapid and simple diagnostic test that can accurately differentiate a deadly P. falciparum infection from a pan-malarial infection caused by P. vivax, P. ovale, or P. malariae

  • For differential diagnosis of a malaria infection
  • Just three simple steps, using one reagent
  • Reliable results in 15 minutes
  • CLIA Complexity: Moderate
  • CPT Code: 87899
  • Sensitivity: 99.7% (P.f.); 93.5% (P.v.)
  • Specificity: 94.2% (P.f.); 99.8% (P.v.)
BNX665000: Malaria Test Kit - Global
BNX665025: Malaria Test Kit - US
BNX665010: Positive Control
Spécifications

Identifiants chimiques

87899
P. vivax Antigen, P. ovale Antigen, P. malariae Antigen
15 min.
Moderate
Whole Blood

Spécifications

87899
P. vivax Antigen, P. ovale Antigen, P. malariae Antigen
BinaxNOW Positive Control
10 Tests
Moderate
Whole Blood
BinaxNOW Positive Control
15 min.
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions