missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abbott™ BinaxNOW™ Malaria Tests
Rapid malaria test cleared by the FDA
Supplier: Abbott™ 660000
Description
A rapid and simple diagnostic test that can accurately differentiate a deadly P. falciparum infection from a pan-malarial infection caused by P. vivax, P. ovale, or P. malariae
- For differential diagnosis of a malaria infection
- Just three simple steps, using one reagent
- Reliable results in 15 minutes
- CLIA Complexity: Moderate
- CPT Code: 87899
- Sensitivity: 99.7% (P.f.); 93.5% (P.v.)
- Specificity: 94.2% (P.f.); 99.8% (P.v.)
BNX665025: Malaria Test Kit - US
BNX665010: Positive Control
Chemical Identifiers
| 87899 | |
| P. vivax Antigen, P. ovale Antigen, P. malariae Antigen | |
| 15 min. |
| Moderate | |
| Whole Blood |
Specifications
| 87899 | |
| P. vivax Antigen, P. ovale Antigen, P. malariae Antigen | |
| Malaria Test Kit (Global) | |
| 15 min. |
| Moderate | |
| Whole Blood | |
| Malaria Test Kit (Global), with built-in control |