missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Bicine, ULTROL™ Grade, Calbiochem™,

A zwitterionic amino acid buffer active in the pH 7.6 to 9.0 range

$1160.00 - $1160.00

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
Synonym N,N-bis-(2-Hydroxyethyl)glycine
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name sodium 2-[bis(2-hydroxyethyl)amino]acetate
SMILES [Na+].OCCN(CCO)CC([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
3913361KG
View Documents
MilliporeSigma™
3913361KG
1 kg
Each for $1,160.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

150-25-4
185.16
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
8761
sodium 2-[bis(2-hydroxyethyl)amino]acetate
C6H12NNaO4
MFCD00004295
N,N-bis-(2-Hydroxyethyl)glycine
CHEBI:40957
[Na+].OCCN(CCO)CC([O-])=O

Specifications

5%, H2O, 250nm
1 kg
C6H12NNaO4
N,N-bis-(2-Hydroxyethyl)glycine
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
sodium 2-[bis(2-hydroxyethyl)amino]acetate
8761
163.2
zwitterionic amino acid buffer
150-25-4
≥99.0%
MFCD00004295
Easily soluble in cold water
[Na+].OCCN(CCO)CC([O-])=O
185.16
CHEBI:40957
by titration
Videos
Safety and Handling

Safety and Handling

Recommended Storage : 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).

SDS
missing translation for 'documents'

missing translation for 'documents'