missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals BICINE, Electrophoresis Grade, 99+%

CAS: 150-25-4 | C6H12NNaO4 | 185.16 g/mol

$104.67 - $252.44

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
Synonym N,N-Bis(2-hydroxyethyl)glycine
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name 2-[bis(2-hydroxyethyl)amino]acetic acid
SMILES [Na+].OCCN(CCO)CC([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6708714
View Documents
Thermo Scientific Chemicals
J6708714
25 g
Each for $104.67
Only null left
 
AAJ6708722
View Documents
Thermo Scientific Chemicals
J6708722
100 g
Each for $252.44
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

150-25-4
185.16
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
8761
2-[bis(2-hydroxyethyl)amino]acetic acid
C6H12NNaO4
MFCD00004295
N,N-Bis(2-hydroxyethyl)glycine
CHEBI:40957
[Na+].OCCN(CCO)CC([O-])=O

Specifications

∼191°C (decomposition)
White
25 g
MFCD00004295
14,1201
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
2-[bis(2-hydroxyethyl)amino]acetic acid
8761
163.17
Electrophoresis
150-25-4
Powder
C6H12NNaO4
1769362
N,N-Bis(2-hydroxyethyl)glycine
[Na+].OCCN(CCO)CC([O-])=O
185.16
CHEBI:40957
≥99%
BICINE
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-755-1

RTECSNumber : MB9700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only