missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Bicine, 99+%, for biochemistry
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
Specifications
Specifications
| Absorbance | 0.05 max. (0.5M, water) at 260nm, 0.05 max. (0.5M, water) at 280nm |
| Melting Point | 190.0°C to 192.0°C |
| CAS Min % | 99.0 |
| Color | White |
| CAS Max % | 100.0 |
| Physical Form | Powder |
| Quantity | 25 g |
| Assay Percent Range | 99+% |
| Infrared Spectrum | Authentic |
| Linear Formula | (HOCH2CH2)2NCH2CO2H |
| Show More |
RUO – Research Use Only