missing translation for 'onlineSavingsMsg'
Learn More

Bicine, 99+%, for analysis

CAS: 150-25-4 | C6H12NNaO4 | 185.16 g/mol

$170.07 - $340.43

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
Synonym N, N-Bis(2-hydroxyethyl)glycine, Diethylolglycine
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name 2-[bis(2-hydroxyethyl)amino]acetic acid
SMILES [Na+].OCCN(CCO)CC([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC172651000
SDS View Documents
Thermo Scientific Chemicals
172651000
100 g Glass bottle
Each for $170.07
Only null left
 
AC172652500
SDS View Documents
Thermo Scientific Chemicals
172652500
250 g Glass bottle
Each for $340.43
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Chemical Identifiers

150-25-4
185.16
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
8761
2-[bis(2-hydroxyethyl)amino]acetic acid
C6H12NNaO4
MFCD00004295
N, N-Bis(2-hydroxyethyl)glycine, Diethylolglycine
CHEBI:40957
[Na+].OCCN(CCO)CC([O-])=O

Specifications

0.05 max. (5% aqueous soln.) at 250nm
150-25-4
100.0
4.0 to 5.5 (10% soln. at 20°C)
100 g
Authentic
(HOCH2CH2)2NCH2CO2H
Glass bottle
N, N-Bis(2-hydroxyethyl)glycine, Diethylolglycine
[Na+].OCCN(CCO)CC([O-])=O
185.16
CHEBI:40957
≥99.0% (Total base)
Bicine, 99+%
(5% aq. soln.) (1cm cell)
99.0
White
Crystalline Powder
99+%
C6H12NNaO4
MFCD00004295
15, 1205
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
2-[bis(2-hydroxyethyl)amino]acetic acid
8761
163.17
Analytical
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-755-1

RTECSNumber : MB9700000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only