missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals BICINE, 99%

CAS: 150-25-4 | C6H12NNaO4 | 185.16 g/mol

$82.62 - $834.99

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name 2-[bis(2-hydroxyethyl)amino]acetic acid
SMILES [Na+].OCCN(CCO)CC([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1495718
View Documents
Thermo Scientific Chemicals
A1495718
50 g
Each for $82.62
Only null left
 
AAA1495730
View Documents
Thermo Scientific Chemicals
A1495730
250 g
Each for $285.99
Only null left
 
AAA149570B
View Documents
Thermo Scientific Chemicals
A149570B
1000 g
Each for $834.99
Only null left
 
Description

Description

Bicine is widely utilized as a buffering agent for low temperature work. It is used in kinetic study of a quaternary transition-state analogue complex of creatine kinase reaction. It is employed in the thin layer ion exchange chromatography method for protein resolution. It is employed in serum guanase determination by the preparation of stable substrate solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bicine is widely utilized as a buffering agent for low temperature work. It is used in kinetic study of a quaternary transition-state analogue complex of creatine kinase reaction. It is employed in the thin layer ion exchange chromatography method for protein resolution. It is employed in serum guanase determination by the preparation of stable substrate solution.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

150-25-4
185.16
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
CHEBI:40957
[Na+].OCCN(CCO)CC([O-])=O
C6H12NNaO4
MFCD00004295
8761
2-[bis(2-hydroxyethyl)amino]acetic acid

Specifications

∽191°C (decomposition)
50 g
C6H12NNaO4
1769362
Soluble in water.
[Na+].OCCN(CCO)CC([O-])=O
185.16
CHEBI:40957
99%
150-25-4
99%
MFCD00004295
14,1201
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
2-[bis(2-hydroxyethyl)amino]acetic acid
8761
163.17
BICINE
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-755-1

RTECSNumber : MB9700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only