Learn More
Thermo Scientific Chemicals BICINE, 99%
Description
Bicine is widely utilized as a buffering agent for low temperature work. It is used in kinetic study of a quaternary transition-state analogue complex of creatine kinase reaction. It is employed in the thin layer ion exchange chromatography method for protein resolution. It is employed in serum guanase determination by the preparation of stable substrate solution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Bicine is widely utilized as a buffering agent for low temperature work. It is used in kinetic study of a quaternary transition-state analogue complex of creatine kinase reaction. It is employed in the thin layer ion exchange chromatography method for protein resolution. It is employed in serum guanase determination by the preparation of stable substrate solution.
Solubility
Soluble in water.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | ∽191°C (decomposition) |
| Quantity | 50 g |
| Assay Percent Range | 99% |
| Beilstein | 1769362 |
| Merck Index | 14,1201 |
| Solubility Information | Soluble in water. |
| Formula Weight | 163.17 |
| Percent Purity | 99% |
| Chemical Name or Material | BICINE |
Safety and Handling
| EINECSNumber | 205-755-1 |
| RTECSNumber | MB9700000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only