missing translation for 'onlineSavingsMsg'
Learn More

BHT, Granular, FCC, 99%, Spectrum™ Chemical

B1095, 128-37-0, C15H24O

Supplier:  Spectrum Chemical Mfg Cor B109525KG

 View more versions of this product

Catalog No. 18600976


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical BHT, Granular, FCC is used in small quantities as a food additive as an antioxidant. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

128-37-0
0.002%
C15H24O
NLZUEZXRPGMBCV-UHFFFAOYSA-N
2,6-di-tert-butyl-4-methylphenol
99%
Crystalline Powder or Lumps
100%
Poly Pail
2.5 kg
CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
220.36
FCC

Chemical Identifiers

128-37-0
220.36
2,6-di-tert-butyl-4-methylphenol
C15H24O
NLZUEZXRPGMBCV-UHFFFAOYSA-N
CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions