missing translation for 'onlineSavingsMsg'
Learn More

BHT, Granular, FCC, 99%, Spectrum™ Chemical

Catalog No. p-7157035
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600974 125 g Amber Glass Bottle
18600975 500 g Amber Glass Bottle
18600976 2.5 kg Poly Pail
18600977 12 kg Fiber Drum
18600978 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18600974 Supplier Spectrum Chemical Mfg Cor Supplier No. B1095125GM
Only null left

B1095, 128-37-0, C15H24O

Spectrum™ Chemical BHT, Granular, FCC is used in small quantities as a food additive as an antioxidant. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 128-37-0
Molecular Formula C15H24O
Molecular Weight (g/mol) 220.36
InChI Key NLZUEZXRPGMBCV-UHFFFAOYSA-N
IUPAC Name 2,6-di-tert-butyl-4-methylphenol
SMILES CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C

Specifications

CAS Max % 100%
Ignition Residue 0.002%
Packaging Amber Glass Bottle
Quantity 125 g
Percent Purity 99%
Grade FCC
Physical Form Crystalline Powder or Lumps