missing translation for 'onlineSavingsMsg'
Learn More

BHA, FCC, 98.5%, Spectrum™ Chemical

BH104, 25013-16-5, C11H16O2

Supplier:  Spectrum Chemical Mfg Cor BH10425KG

 View more versions of this product

Catalog No. 18600971

Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical BHA, FCC is an antioxidant and preservative used in food, food packaging and animal feed. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

25013-16-5
0.05%
C11H16O2
MRBKEAMVRSLQPH-UHFFFAOYSA-N
2-tert-butyl-4-methoxyphenol
98.5%
Crystals or Flakes
100%
Amber Glass Bottle
2.5 kg
COC1=CC=C(O)C(=C1)C(C)(C)C
180.25
FCC

Chemical Identifiers

25013-16-5
180.25
2-tert-butyl-4-methoxyphenol
C11H16O2
MRBKEAMVRSLQPH-UHFFFAOYSA-N
COC1=CC=C(O)C(=C1)C(C)(C)C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions