missing translation for 'onlineSavingsMsg'
Learn More

BHA, FCC, 98.5%, Spectrum™ Chemical

Catalog No. p-7157091
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600969 125 g Amber Glass Bottle
18600970 500 g Amber Glass Bottle
18600971 2.5 kg Amber Glass Bottle
18600972 12 kg Amber Glass Bottle
18600973 45 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18600969 Supplier Spectrum Chemical Mfg Cor Supplier No. BH104125GM
Only null left

BH104, 25013-16-5, C11H16O2

Spectrum™ Chemical BHA, FCC is an antioxidant and preservative used in food, food packaging and animal feed. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 25013-16-5
Molecular Formula C11H16O2
Molecular Weight (g/mol) 180.25
InChI Key MRBKEAMVRSLQPH-UHFFFAOYSA-N
IUPAC Name 2-tert-butyl-4-methoxyphenol
SMILES COC1=CC=C(O)C(=C1)C(C)(C)C

Specifications

CAS Max % 100%
Ignition Residue 0.05%
Packaging Amber Glass Bottle
Quantity 125 g
Percent Purity 98.5%
Grade FCC
Physical Form Crystals or Flakes