Learn More
Bezafibrate, 98+%
A peroxisome proliferator-activated receptor agonist and hypolipidemic agent | CAS: 41859-67-0 | C19H20ClNO4 | 361.82 g/mol
Supplier: Thermo Scientific Chemicals J6141203
Description
A peroxisome proliferator-activated receptor agonist and hypolipidemic agent
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Bezafibrate | |
| 183°C to 186°C | |
| C19H20ClNO4 | |
| 1 g | |
| bezafibrate, bezalip, cedur, bezafibrat, befizal, bezafibrato, bezafibratum, sklerofibrat, azufibrat, difaterol | |
| CC(C)(OC1=CC=C(CCNC(=O)C2=CC=C(Cl)C=C2)C=C1)C(O)=O | |
| 361.82 | |
| CHEBI:47612 | |
| ≥98% |
| 41859-67-0 | |
| White | |
| MFCD00078970 | |
| 14,1195 | |
| IIBYAHWJQTYFKB-UHFFFAOYSA-N | |
| 2-(4-{2-[(4-chlorophenyl)formamido]ethyl}phenoxy)-2-methylpropanoic acid | |
| 39042 | |
| 361.82 | |
| Powder |
Chemical Identifiers
| 41859-67-0 | |
| 361.82 | |
| IIBYAHWJQTYFKB-UHFFFAOYSA-N | |
| 39042 | |
| 2-(4-{2-[(4-chlorophenyl)formamido]ethyl}phenoxy)-2-methylpropanoic acid |
| C19H20ClNO4 | |
| MFCD00078970 | |
| bezafibrate, bezalip, cedur, bezafibrat, befizal, bezafibrato, bezafibratum, sklerofibrat, azufibrat, difaterol | |
| CHEBI:47612 | |
| CC(C)(OC1=CC=C(CCNC(=O)C2=CC=C(Cl)C=C2)C=C1)C(O)=O |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 255-567-9
RTECSNumber : UE8755000
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only