missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Bexarotene

Catalog No. 581910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
581910 10 mg
581950 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 581910 Supplier Tocris Bioscience™ Supplier No. 5819/10

Potent and selective RXR agonist

Bexarotene is a potent and selective RXR agonist (EC50 values are 24, 25 and 33 nM for RXRβ, RXRγ and RXRα, respectively). Exhibits >300-fold selectivity for RXR over RAR receptors. Inhibits tumor cell invasion and migration in vitro, and inhibits metastasis and angiogenesis in mouse models. Also promotes brown adipogenic differentiation and induces brown adipogenic reprogramming from myoblasts.

Specifications

Chemical Name or Material 4-[1-(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)ethenyl]benzoic acid
CAS 153559-49-0
Synonym LGD 1069
Quantity 10 mg
Target Retinoid X Receptor Agonists
Molecular Formula C24H28O2
Formula Weight 348.48
Purity >99%