missing translation for 'onlineSavingsMsg'
Learn More

Bevantolol hydrochloride, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 468872500

 View more versions of this product

Catalog No. AC468872500


Only null left

Description

Description

Specifications

Specifications

Bevantolol hydrochloride
Available
Titration Argentometric
C20H28ClNO4
00941389
225°C to 228°C
Slightly soluble in water; Soluble in methanol and chloroform; Insoluble in ether, ethyl acetate and hexane
[H+].[Cl-].COC1=CC=C(CCNCC(O)COC2=CC=CC(C)=C2)C=C1OC
381.90
381.9
Glass Bottle
42864-78-8
Powder
97.5% to 102.5%
White to Yellow to Beige
1194
FJTKCFSPYUMXJB-UHFFFAOYNA-N
hydrogen 1-{[2-(3,4-dimethoxyphenyl)ethyl]amino}-3-(3-methylphenoxy)propan-2-ol chloride
250 mg

Chemical Identifiers

42864-78-8
381.90
FJTKCFSPYUMXJB-UHFFFAOYNA-N
[H+].[Cl-].COC1=CC=C(CCNCC(O)COC2=CC=CC(C)=C2)C=C1OC
C20H28ClNO4
00941389
hydrogen 1-{[2-(3,4-dimethoxyphenyl)ethyl]amino}-3-(3-methylphenoxy)propan-2-ol chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS07: Exclamation mark

ShelfLife : 5 years

RTECSNumber : UB2709000

Recommended Storage : Refrigerator +4°C

SDS
Documents

Documents

Promotions

Promotions