missing translation for 'onlineSavingsMsg'
Learn More

Bevantolol hydrochloride, Thermo Scientific™

Catalog No. p-7232420
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468872500 250 mg
AC468870010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468872500 Supplier Thermo Scientific Chemicals Supplier No. 468872500
Only null left

Chemical Identifiers

CAS 42864-78-8
Molecular Formula C20H28ClNO4
Molecular Weight (g/mol) 381.90
MDL Number 00941389
InChI Key FJTKCFSPYUMXJB-UHFFFAOYNA-N
IUPAC Name hydrogen 1-{[2-(3,4-dimethoxyphenyl)ethyl]amino}-3-(3-methylphenoxy)propan-2-ol chloride
SMILES [H+].[Cl-].COC1=CC=C(CCNCC(O)COC2=CC=CC(C)=C2)C=C1OC

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Concentration or Composition Notes Titration Argentometric
Physical Form Powder
Concentration 97.5% to 102.5%
Color White to Yellow to Beige
Melting Point 225°C to 228°C
Merck Index 1194
Solubility Slightly soluble in water; Soluble in methanol and chloroform; Insoluble in ether, ethyl acetate and hexane
Quantity 250 mg
Formula Weight 381.9
Chemical Name or Material Bevantolol hydrochloride
Show More Show Less
Hazard Category GHS07: Exclamation mark
ShelfLife 5 years
RTECSNumber UB2709000
Recommended Storage Refrigerator +4°C