missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Betulin

Catalog No. AC466550010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466550010 1 g
AC466550050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466550010 Supplier Thermo Scientific Chemicals Supplier No. 466550010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 473-98-3 | C30H50O2, C30H50O2 | 442.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 473-98-3
Molecular Formula C30H50O2

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: .. Other solubilities: soluble in acetic acid
Melting Point 256.0°C to 257.0°C
Formula Weight 442.72
InChI Key FVWJYYTZTCVBKE-ROUWMTJPSA-N
SMILES CC(=C)[C@@H]1CC[C@]2(CO)CC[C@]3(C)[C@H](CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]12
IUPAC Name (1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-3a-(hydroxymethyl)-5a,5b,8,8,11a-pentamethyl-1-(prop-1-en-2-yl)-icosahydro-1H-cyclopenta[a]chrysen-9-ol
Molecular Weight (g/mol) 442.73
Chemical Name or Material Betulin
EINECSNumber 207-475-5

RUO – Research Use Only