missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Betaxolol Hydrochloride 98.0+%, TCI America™
Supplier: TCI America B4474250MG
Specifications
| Betaxolol Hydrochloride | |
| 115°C | |
| C18H30ClNO3 | |
| 250 mg | |
| CHDPSNLJFOQTRK-UHFFFAOYSA-N | |
| 1-[4-[2-(cyclopropylmethoxy)ethyl]phenoxy]-3-(propan-2-ylamino)propan-2-ol;hydrochloride | |
| 107952 | |
| 343.89 | |
| Crystalline Powder |
| 63659-19-8 | |
| White-Yellow | |
| MFCD00242959 | |
| betaxolol hydrochloride, betaxolol hcl, betoptima, kerlone, betoptic, kerlon, lokren, betaxolo hydrochloride, betoptic s | |
| CC(C)NCC(COC1=CC=C(C=C1)CCOCC2CC2)O.Cl | |
| 343.892 | |
| CHEBI:643228 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 63659-19-8 | |
| 343.892 | |
| CHDPSNLJFOQTRK-UHFFFAOYSA-N | |
| 107952 | |
| 1-[4-[2-(cyclopropylmethoxy)ethyl]phenoxy]-3-(propan-2-ylamino)propan-2-ol;hydrochloride |
| C18H30ClNO3 | |
| MFCD00242959 | |
| betaxolol hydrochloride, betaxolol hcl, betoptima, kerlone, betoptic, kerlon, lokren, betaxolo hydrochloride, betoptic s | |
| CHEBI:643228 | |
| CC(C)NCC(COC1=CC=C(C=C1)CCOCC2CC2)O.Cl |
Safety and Handling
RTECSNumber : UA9823000
TSCA : No