missing translation for 'onlineSavingsMsg'
Learn More

Betamethasone Valerate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013542
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013542 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013542 Supplier Merck Emd Millipore Supplier No. PHR17801G

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 2152-44-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C27H37 FO6
MDL Number MFCD00867446
Synonym Betamethasone 17-valerate; 1,4-Pregnadiene-11 beta,17 alpha,21-triol-9 alpha-fluoro-16 beta-methyl-3,20-dione 17-valerate; 9 alpha-Fluoro-16 beta-methyl-11 beta,17 alpha,21-trihydroxy-1,4-pregnadiene-3,20-dione 17-valerate; 9 alpha-Fluoro-16 beta-methylprednisolone 17-valerate; Betnovate
Type Certified Reference Material
Quantity 1 g
Formula Weight 476.58
Grade Certified Reference Material