accessibility menu, dialog, popup

UserName

CAS RN 2152-44-5

H3COO(R)(S)H3C(S)(S)O(S)H3C(R)F(S)OH(S)H3COHO

CAS RN 2152-44-5

IUPAC Name: (1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-10-hydroxy-1-(2-hydroxyacetyl)-2,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl pentanoate
Synonyms: betamethasone valerate betamethasone 17-valerate betamethasone valerate (usan)(jan)
Molecular Weight (g/mol): 476.59
Molecular Formula: C27H37FO6
InChi Key: SNHRLVCMMWUAJD-SUYDQAKGSA-N
SMILES: CCCCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CO
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results