missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BetaBasic™ CN HPLC Columns, 5μm Particle Size

Catalog No. p-4523339
Change view
Click to view available options
Length (Metric):
50 mm
250 mm
Diameter (Metric):
2.1 mm
4.6 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
71705052130 50 mm 2.1 mm
71705254630 250 mm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71705052130 Supplier Thermo Scientific™ Supplier No. 71705052130
Only null left

Thermo ScientificTM BetaBasicTM CN HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.

  • Pore size suitable for small molecules, peptides and protein digests
  • Superb pH stability

Ordering Information:

Other column dimensions are also available. Please call Customer Service for more information.

Specifications

Column Format Analytical Column
Diameter (Metric) 2.1 mm
For Use With LC/MS, APCI
Length (Metric) 50 mm
Particle Size 5 μm
pH 2 to 9
Pore Size 150 Å
Surface Area 200 m²/g
USP Type L10
Stationary Phase Cyano (CN)
Product Line Beta
Packing Material Silica
Carbon Load 5%
Column Type Mixed Mode
Endcapped Yes
Phase Normal and Reversed Phase
Show More Show Less