missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BetaBasic™ CN HPLC Columns, 5μm Particle Size
Thermo ScientificTM BetaBasicTM CN HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.
Supplier: Thermo Scientific™ 71705254630
Description
Thermo ScientificTM BetaBasicTM CN HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.
- Pore size suitable for small molecules, peptides and protein digests
- Superb pH stability
Ordering Information:
Other column dimensions are also available. Please call Customer Service for more information.
Specifications
| Analytical Column | |
| LC/MS, APCI | |
| 5 μm | |
| 150 Å | |
| L10 | |
| Beta | |
| 5% | |
| Yes |
| 4.6 mm | |
| 250 mm | |
| 2 to 9 | |
| 200 m²/g | |
| Cyano (CN) | |
| Silica | |
| Mixed Mode | |
| Normal and Reversed Phase |