missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BetaBasic™ 8 Guard Cartridges, 5μm Particle Size
Thermo Scientific™ BetaBasic™ 8 Guard Cartridges are available in limited dimensions. Please contact Customer Service for more details.
$1205.51 - $1205.51
Specifications
| Carbon Load | 7% |
|---|---|
| pH | 2 to 9 |
| Particle Size | 5 μm |
| Length (Metric) | 10 mm |
| Pore Size | 150Å |
| Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Particle Size | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Particle Size | Price | Quantity | |||
|
71405014001
|
Thermo Scientific™
71405014001 |
4 mm | 10 mm | 5 μm |
Pack of 4 Each for $1,205.51
|
|
|||
Description
Thermo Scientific™ BetaBasic™ 8 Guard Cartridges are available in limited dimensions. Please contact Customer Service for more details.
- The 10mm design offers maximum protection with minimal increase in retention
- Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
- Highly efficient and reproducible Thermo Scientific columns
- 150Å pore size for small molecules, peptides and protein digests
- Superb pH stability
Specifications
| 7% | |
| 5 μm | |
| 150Å | |
| Uniguard Direct-connection and Stand Alone Holder | |
| L7 |
| 2 to 9 | |
| 10 mm | |
| 200 m2/g | |
| Yes |