missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BetaBasic™ 18 Javelin™ Guard Columns

Catalog No. 71505014006
Change view
Click to view available options
Diameter (Metric) Inner:
2.1 mm
4 mm
Length (Metric):
10 mm
20 mm
Particle Size:
5 μm
Catalog No. Diameter (Metric) Inner Length (Metric) Particle Size
71505014006 4 mm 10 mm 5 μm
71505022106 2.1 mm 20 mm 5 μm
2 options

Catalog No. 71505014006

Supplier: Thermo Scientific™ 71505014006

Only null left

Offer convenient, easy to use protection for analytical HPLC columns

  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
  • Highly efficient and reproducible Thermo Scientific columns
  • 150Å pore size for small molecules, peptides and protein digests
  • Superb pH stability

Order Info

Javelin guards are available in other Thermo Scientific phases. Please call your Customer Service Representative for more information.

Specifications

Particle Size 5 μm
Length (Metric) 10 mm
Diameter (Metric) Inner 4 mm
Stationary Phase BetaBasic 18
Product Line Beta