missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Nicotinamide adenine dinucleotide, disodium salt, hydrate, 95+%, reduced form
CAS: 1949720-50-6 | C21H27N7Na2O14P2 | 709.41 g/mol
$299.15 - $1092.72
Chemical Identifiers
| CAS | 1949720-50-6 |
|---|---|
| Molecular Formula | C21H27N7Na2O14P2 |
| Molecular Weight (g/mol) | 709.41 |
| MDL Number | MFCD00036200 |
| InChI Key | QRGNQKGQENGQSE-WUEGHLCSSA-L |
| Synonym | beta-nadh disodium salt |
| PubChem CID | 131673989 |
| IUPAC Name | [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium |
| SMILES | [Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC271100010
|
Thermo Scientific Chemicals
271100010 |
1 g | Glass bottle |
Each for $299.15
|
|
||||
|
AC271100050
|
Thermo Scientific Chemicals
271100050 |
5 g | Glass bottle |
Each for $1,092.72
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1949720-50-6 | |
| 709.41 | |
| QRGNQKGQENGQSE-WUEGHLCSSA-L | |
| 131673989 | |
| [Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O |
| C21H27N7Na2O14P2 | |
| MFCD00036200 | |
| beta-nadh disodium salt | |
| [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium |
Specifications
| 1949720-50-6 | |
| 0.50mg/mL NADH in 10mM TRIS solution | |
| 140.0°C to 142.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00036200 | |
| beta-nadh disodium salt | |
| QRGNQKGQENGQSE-WUEGHLCSSA-L | |
| [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium | |
| 131673989 | |
| 95+% | |
| β-Nicotinamide adenine dinucleotide, disodium salt, hydrate, Reduced Form |
| 95.0 | |
| 100.0 | |
| Yellow | |
| 95% min. (HPLC) | |
| C21H27N7Na2O14P2 | |
| 1 g | |
| Solubility in water: soluble | |
| [Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O | |
| 709.41 | |
| 709.4 | |
| Powder |
RUO – Research Use Only