missing translation for 'onlineSavingsMsg'
Learn More

beta-Nicotinamide adenine dinucleotide, disodium salt, hydrate, 95+%, reduced form

CAS: 1949720-50-6 | C21H27N7Na2O14P2 | 709.41 g/mol

Supplier:  Thermo Scientific Chemicals 271100050

 View more versions of this product

Catalog No. AC271100050


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

β-Nicotinamide adenine dinucleotide, disodium salt, hydrate
1949720-50-6
0.50mg/mL NADH in 10mM TRIS solution
140.0°C to 142.0°C
Authentic
Glass bottle
MFCD00036200
beta-nadh disodium salt
QRGNQKGQENGQSE-WUEGHLCSSA-L
[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium
131673989
95+%
Reduced Form
95.0
100.0
Yellow
95% min. (HPLC)
C21H27N7Na2O14P2
5 g
Solubility in water: soluble
[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
709.41
709.4
Powder

Chemical Identifiers

1949720-50-6
709.41
QRGNQKGQENGQSE-WUEGHLCSSA-L
131673989
[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
C21H27N7Na2O14P2
MFCD00036200
beta-nadh disodium salt
[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only