missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Nicotinamide adenine dinucleotide, disodium salt, hydrate, 95+%, reduced form
CAS: 1949720-50-6 | C21H27N7Na2O14P2 | 709.41 g/mol
Supplier: Thermo Scientific Chemicals 271100010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| β-Nicotinamide adenine dinucleotide, disodium salt, hydrate | |
| 1949720-50-6 | |
| 0.50mg/mL NADH in 10mM TRIS solution | |
| 140.0°C to 142.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00036200 | |
| beta-nadh disodium salt | |
| QRGNQKGQENGQSE-WUEGHLCSSA-L | |
| [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium | |
| 131673989 | |
| 95+% |
| Reduced Form | |
| 95.0 | |
| 100.0 | |
| Yellow | |
| 95% min. (HPLC) | |
| C21H27N7Na2O14P2 | |
| 1 g | |
| Solubility in water: soluble | |
| [Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O | |
| 709.41 | |
| 709.4 | |
| Powder |
Chemical Identifiers
| 1949720-50-6 | |
| 709.41 | |
| QRGNQKGQENGQSE-WUEGHLCSSA-L | |
| 131673989 | |
| [Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O |
| C21H27N7Na2O14P2 | |
| MFCD00036200 | |
| beta-nadh disodium salt | |
| [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium |
RUO – Research Use Only