missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Beta-Glucuronidase

For analysis of steroid-gluceronides

$151.64 - $917.46

Specifications

Concentration beta-Glucuronidase: >50,000 U/mL, Sulfatase: 5,000 U/mL
Color Brown
For Use With (Application) Enzymatic hydrolysis of Glucuronides from Serum Plasma and other fluids prior to analysis by Enzyme immunoassay, mass spectrometry, GC, HPLC
Content And Storage 4°C
Source Helix pomatia
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN15228401
View Documents
MP Biomedicals™
0215228401
1 mL
Each for $151.64
Only null left
 
ICN15228410
View Documents
MP Biomedicals™
0215228410
10 mL
Each for $917.46
Only null left
 
Description

Description

Beta-Glucuronidase, [9001-45-0]

  • For analysis of steroid-gluceronides.
  • In 65% saturated ammonium sulfate
  • Activity: ≤5000 u/mL
  • Unit Definition: Amount that liberates 1.0μg of phenolphthalein from phenolphthalein glucuronide per hour at pH 5.0 and 37°C.

Specifications

Specifications

beta-Glucuronidase: >50,000 U/mL, Sulfatase: 5,000 U/mL
Enzymatic hydrolysis of Glucuronides from Serum Plasma and other fluids prior to analysis by Enzyme immunoassay, mass spectrometry, GC, HPLC
Helix pomatia
D-glucuronic Acid, D-galacturonic Acid and D-glucaro-1,4-lactone
Brown
4°C
beta-Glucuronidase
Solution
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'