missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Citronellol 95.0+%, TCI America™
$51.26 - $237.33
Chemical Identifiers
| CAS | 106-22-9 |
|---|---|
| Molecular Formula | C10H20O |
| Molecular Weight (g/mol) | 156.269 |
| MDL Number | MFCD00002935 |
| InChI Key | QMVPMAAFGQKVCJ-UHFFFAOYSA-N |
| Synonym | citronellol, beta-citronellol, 3,7-dimethyl-6-octen-1-ol, dl-citronellol, cephrol, elenol, 6-octen-1-ol, 3,7-dimethyl, rodinol, 2,3-dihydrogeraniol, 2,6-dimethyl-2-octen-8-ol |
| PubChem CID | 8842 |
| ChEBI | CHEBI:50462 |
| IUPAC Name | 3,7-dimethyloct-6-en-1-ol |
| SMILES | CC(CCC=C(C)C)CCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
C037025ML
|
N/A | 25 mL |
Each for $51.26
|
|
|||||
|
C0370100ML
|
N/A | 100 mL |
Each for $96.84
|
|
|||||
|
C0370500ML
|
N/A | 500 mL |
Each for $237.33
|
|
|||||
Chemical Identifiers
| C10H20O | |
| MFCD00002935 | |
| citronellol, beta-citronellol, 3,7-dimethyl-6-octen-1-ol, dl-citronellol, cephrol, elenol, 6-octen-1-ol, 3,7-dimethyl, rodinol, 2,3-dihydrogeraniol, 2,6-dimethyl-2-octen-8-ol | |
| CHEBI:50462 | |
| CC(CCC=C(C)C)CCO |
Specifications
| 106-22-9 | |
| 220°C | |
| MFCD00002935 | |
| QMVPMAAFGQKVCJ-UHFFFAOYSA-N | |
| 3,7-dimethyloct-6-en-1-ol | |
| 8842 | |
| 156.27 | |
| Liquid |
| Yellow | |
| C10H20O | |
| citronellol, beta-citronellol, 3,7-dimethyl-6-octen-1-ol, dl-citronellol, cephrol, elenol, 6-octen-1-ol, 3,7-dimethyl, rodinol, 2,3-dihydrogeraniol, 2,6-dimethyl-2-octen-8-ol | |
| CC(CCC=C(C)C)CCO | |
| 156.269 | |
| CHEBI:50462 | |
| ≥95.0% (GC) | |
| beta-Citronellol |
Safety and Handling
EINECSNumber : (2)-0258
RTECSNumber : RH3400000
TSCA : Yes