missing translation for 'onlineSavingsMsg'
Learn More

beta-Citronellol 95.0+%, TCI America™

Catalog No. C0370100ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0370100ML 100 mL
C037025ML 25 mL
C0370500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0370100ML Supplier TCI America Supplier No. C0370100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106-22-9
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00002935
InChI Key QMVPMAAFGQKVCJ-UHFFFAOYSA-N
Synonym citronellol, beta-citronellol, 3,7-dimethyl-6-octen-1-ol, dl-citronellol, cephrol, elenol, 6-octen-1-ol, 3,7-dimethyl, rodinol, 2,3-dihydrogeraniol, 2,6-dimethyl-2-octen-8-ol
PubChem CID 8842
ChEBI CHEBI:50462
IUPAC Name 3,7-dimethyloct-6-en-1-ol
SMILES CC(CCC=C(C)C)CCO

Specifications

Color Yellow
Boiling Point 220°C
Quantity 100 mL
Formula Weight 156.27
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material beta-Citronellol
EINECSNumber (2)-0258
RTECSNumber RH3400000
TSCA Yes