missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Bromo-4-nitrophenetole 97.0+%, TCI America™
Supplier: TCI America B32545G
Specifications
| beta-Bromo-4-nitrophenetole | |
| 66°C | |
| 170°C | |
| MFCD00031363 | |
| 1-2-bromoethoxy-4-nitrobenzene, 2-bromoethyl 4-nitrophenyl ether, beta-bromo-4-nitrophenetole, 2-4-nitrophenoxy ethyl bromide, 4-2-bromoethoxy-1-nitrobenzene, 1-2-bromoethyloxy-4-nitro-benzene, benzene, 1-2-bromoethoxy-4-nitro, 4-2-bromoethoxy nitrobenzene, acmc-1azdi | |
| [O-][N+](=O)C1=CC=C(OCCBr)C=C1 | |
| 246.06 | |
| 246.06 | |
| Crystalline Powder |
| 13288-06-7 | |
| Yellow | |
| C8H8BrNO3 | |
| 5 g | |
| YQWCBDNNEZHPMA-UHFFFAOYSA-N | |
| 1-(2-bromoethoxy)-4-nitrobenzene | |
| 235987 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 13288-06-7 | |
| 246.06 | |
| YQWCBDNNEZHPMA-UHFFFAOYSA-N | |
| 235987 | |
| [O-][N+](=O)C1=CC=C(OCCBr)C=C1 |
| C8H8BrNO3 | |
| MFCD00031363 | |
| 1-2-bromoethoxy-4-nitrobenzene, 2-bromoethyl 4-nitrophenyl ether, beta-bromo-4-nitrophenetole, 2-4-nitrophenoxy ethyl bromide, 4-2-bromoethoxy-1-nitrobenzene, 1-2-bromoethyloxy-4-nitro-benzene, benzene, 1-2-bromoethoxy-4-nitro, 4-2-bromoethoxy nitrobenzene, acmc-1azdi | |
| 1-(2-bromoethoxy)-4-nitrobenzene |
Safety and Handling
TSCA : No